Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Flucofuron, Dr. Ehrenstorfer, LGC Standards
Flucofuron, Dr. Ehrenstorfer, LGC Standards
77678-082
:
LGC Standards
New Product
:
Flucofuron, Dr. Ehrenstorfer, LGC Standards
77678-082
:
LGC Standards
:
DRE-C13697000
New Product
:
CAS No.:
370-50-3
Description:
Flucofuron
Size:
10 mg
Formula:
C₁₅H₈Cl₂F₆N₂O
Molecular Weight:
417.1332 g/mol
Cat. No.:
77678-082