Fast Green FCF, Dr. Ehrenstorfer, LGC Standards
Catalog # 77678-015
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:2353-45-9
- Description:Fast Green FCF
- Size:100 mg
- Formula:C₃₇H₃₄N₂Na₂O₁₀S₃
- Molecular Weight:808.86 g/mol
- Cat. No.:77678-015