Doxycycline hyclate, Dr. Ehrenstorfer, LGC Standards
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:24390-14-5
- Formula:C₂₂H₂₄N₂O₈·HCl·0.5H₂O·0.5C₂H₆O
- Molecular Weight:512.94 g/mol (503.94 anhy)
Specifications
Frequently Bought Together