Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Digoxin, Dr. Ehrenstorfer, LGC Standards
Digoxin, Dr. Ehrenstorfer, LGC Standards
Digoxin, Dr. Ehrenstorfer, LGC Standards
77677-869
:
LGC Standards
New Product
:
Digoxin, Dr. Ehrenstorfer, LGC Standards
77677-869
:
LGC Standards
:
DRE-C12633850
New Product
:
CAS No.:
20830-75-5
Description:
Digoxin
Size:
100 mg
Formula:
C₄₁H₆₄O₁₄
Molecular Weight:
780.95 g/mol
Cat. No.:
77677-869