Difloxacin hydrochloride, Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-864
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:91296-86-5
- Description:Difloxacin hydrochloride
- Size:100 mg
- Formula:C₂₁H₁₉F₂N₃O₃·HCl
- Molecular Weight:435.85 g/mol
- Cat. No.:77677-864