1-(2,4-Dichlorophenyl)-2-imidazol-1-ylethanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-838
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:24155-42-8
- Description:1-(2,4-Dichlorophenyl)-2-imidazol-1-ylethanol
- Size:100 mg
- Formula:C₁₁H₁₀Cl₂N₂O
- Molecular Weight:257.11 g/mol
- Cat. No.:77677-838