Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
2,6-Di-tert-butyl-4-methylphenol, Dr. Ehrenstorfer, LGC Standards
2,6-Di-tert-butyl-4-methylphenol, Dr. Ehrenstorfer, LGC Standards
2,6-Di-tert-butyl-4-methylphenol, Dr. Ehrenstorfer, LGC Standards
77677-816
:
LGC Standards
New Product
:
2,6-Di-tert-butyl-4-methylphenol, Dr. Ehrenstorfer, LGC Standards
77677-816
:
LGC Standards
:
DRE-C12253500
New Product
:
CAS No.:
128-37-0
Description:
2,6-Di-tert-butyl-4-methylphenol
Size:
250 mg
Formula:
C₁₅H₂₄O
Molecular Weight:
220.35 g/mol
Cat. No.:
77677-816