Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
lambda-Cyhalothrin, Dr. Ehrenstorfer, LGC Standards
lambda-Cyhalothrin, Dr. Ehrenstorfer, LGC Standards
lambda-Cyhalothrin, Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-760
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
lambda-Cyhalothrin, Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-760
Supplier:
LGC Standards
Supplier Number:
DRE-C11860000
New Product
CAS Number:
Specifications
CAS No.:
91465-08-6
Description:
lambda-Cyhalothrin
Size:
100 mg
Formula:
C₂₃H₁₉ClF₃NO₃
Cat. No.:
77677-760
Specifications
About this item
Product Details
Product Details