1,2-Cyclohexanedicarboxylic acid, bis(2-ethylhexyl) ester, Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-745
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:84-71-9
- Description:1,2-Cyclohexanedicarboxylic acid, bis(2-ethylhexyl) ester
- Size:100 mg
- Formula:C₂₄H₄₄O₄
- Molecular Weight:396.6 g/mol
- Cat. No.:77677-745