Chlorotetracycline hydrochloride, Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-689
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:64-72-2
- Description:Chlorotetracycline hydrochloride
- Size:250 mg
- Formula:C₂₂H₂₃ClN₂O₈·HCl
- Molecular Weight:515.34 g/mol
- Cat. No.:77677-689