Chlorobenzilate, Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-670
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:510-15-6
- Description:Chlorobenzilate
- Size:100 mg
- Formula:C₁₆H₁₄Cl₂O₃
- Molecular Weight:325.19 g/mol
- Cat. No.:77677-670