$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Basic Red 1, Dr. Ehrenstorfer, LGC Standards
Basic Red 1, Dr. Ehrenstorfer, LGC Standards
77677-515
:
LGC Standards
New Product
:
Basic Red 1, Dr. Ehrenstorfer, LGC Standards
77677-515
:
LGC Standards
:
DRE-C10424950
New Product
:
CAS No.:
989-38-8
Description:
Basic Red 1
Size:
100 mg
Formula:
C₂₈H₃₁ClN₂O₃
Molecular Weight:
479.02 g/mol
Cat. No.:
77677-515