Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Atrazine D5 (ethylamino D5), Dr. Ehrenstorfer, LGC Standards
Atrazine D5 (ethylamino D5), Dr. Ehrenstorfer, LGC Standards
Atrazine D5 (ethylamino D5), Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-495
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Atrazine D5 (ethylamino D5), Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-495
Supplier:
LGC Standards
Supplier Number:
DRE-C10330100
New Product
CAS Number:
Specifications
CAS No.:
163165-75-1
Description:
Atrazine D5 (ethylamino D5)
Size:
10 mg
Cat. No.:
77677-495
Specifications
About this item
Product Details
Product Details