Ancymidol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-477
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:12771-68-5
- Description:Ancymidol
- Size:100 mg
- Formula:C₁₅H₁₆N₂O₂
- Molecular Weight:256.31 g/mol
- Cat. No.:77677-477