Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
4-Aminoquinaldine, Dr. Ehrenstorfer, LGC Standards
4-Aminoquinaldine, Dr. Ehrenstorfer, LGC Standards
77677-470
:
LGC Standards
New Product
:
4-Aminoquinaldine, Dr. Ehrenstorfer, LGC Standards
77677-470
:
LGC Standards
:
DRE-C10225000
New Product
:
CAS No.:
6628-04-2
Description:
4-Aminoquinaldine
Size:
100 mg
Formula:
C₁₀H₁₀N₂
Molecular Weight:
158.2 g/mol
Cat. No.:
77677-470