2-Aminonaphthalene, Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-463
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:91-59-8
- Description:2-Aminonaphthalene
- Size:10 mg
- Formula:C₁₀H₉N
- Molecular Weight:143.19 g/mol
- Cat. No.:77677-463