4-Aminobiphenyl, Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-455
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:92-67-1
- Description:4-Aminobiphenyl
- Size:100 mg
- Formula:C₁₂H₁₁N
- Molecular Weight:169.23 g/mol
- Cat. No.:77677-455