Ambroxol hydrochloride, Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-446
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:23828-92-4
- Description:Ambroxol hydrochloride
- Size:100 mg
- Formula:C₁₃H₁₈Br₂N₂O.HCl
- Molecular Weight:414.57 g/mol
- Cat. No.:77677-446