$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Aclonifen, Dr. Ehrenstorfer, LGC Standards
Aclonifen, Dr. Ehrenstorfer, LGC Standards
77677-424
:
LGC Standards
New Product
:
Aclonifen, Dr. Ehrenstorfer, LGC Standards
77677-424
:
LGC Standards
:
DRE-C10042000
New Product
:
CAS No.:
74070-46-5
Description:
Aclonifen
Size:
250 mg
Formula:
C₁₂H₉ClN₂O₃
Molecular Weight:
264.67 g/mol
Cat. No.:
77677-424