Acetamiprid, Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-403
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:135410-20-7
- Description:Acetamiprid
- Size:100 mg
- Formula:C₁₀H₁₁ClN₄
- Molecular Weight:222.67 g/mol
- Cat. No.:77677-403