Order Entry
Puerto Rico
ContactUsLinkComponent
 
Chloroparaffin C10-C13 63% Cl 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
  77691-687
New Product
 :  
undefined
Chloroparaffin C10-C13 63% Cl 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
  77691-687
 :  DRE-A23106300CY-100
New Product
 :  

 

  • CAS No.:
    85535-84-8
  • Description:
    Chloroparaffin C10-C13 63% Cl 100 µg/mL in Cyclohexane
  • Size:
    1 ml
  • Cat. No.:
    77691-687