Vitamin D3 10 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77693-464
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:67-97-0
- Description:Vitamin D3 10 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₂₇H₄₄O
- Molecular Weight:384.65 g/mol
- Cat. No.:77693-464