Venlafaxine O-desmethyl 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77693-459
New Product
:
- CAS No.:93413-62-8
- Description:Venlafaxine O-desmethyl 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₆H₂₅NO₂
- Molecular Weight:263.38 g/mol
- Cat. No.:77693-459