Vedaprofen 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77693-458
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:71109-09-6
- Description:Vedaprofen 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₉H₂₂O₂
- Molecular Weight:282.38 g/mol
- Cat. No.:77693-458