Tulathromycin A 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77693-452
New Product
:
- CAS No.:217500-96-4
- Description:Tulathromycin A 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₄₁H₇₉N₃O₁₂
- Molecular Weight:806.08 g/mol
- Cat. No.:77693-452