1,1,2-Trifluoro-2-(heptafluoropropoxy)ethene 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-681
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:1623-05-8
- Description:1,1,2-Trifluoro-2-(heptafluoropropoxy)ethene 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₅F₁₀O
- Molecular Weight:266.04 g/mol
- Cat. No.:77691-681