Triclosan-methyl ether 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77693-446
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:4640-01-1
- Description:Triclosan-methyl ether 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₃H₉Cl₃O₂
- Molecular Weight:303.56 g/mol
- Cat. No.:77693-446