$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Trenbolone 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Trenbolone 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77693-442
:
LGC Standards
New Product
:
Trenbolone 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77693-442
:
LGC Standards
:
DRE-A17607700AL-100
New Product
:
CAS No.:
10161-33-8
Description:
Trenbolone 100 µg/mL in Acetonitrile
Size:
1 ml
Formula:
C₁₈H₂₂O₂
Cat. No.:
77693-442