Order Entry
Puerto Rico
ContactUsLinkComponent
 
Tizanidine hydrochloride 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77693-438
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Tizanidine hydrochloride 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77693-438
Supplier:  LGC Standards
Supplier Number:  DRE-A17587600AL-100
New Product
CAS Number:  

Specifications

  • CAS No.:
    64461-82-1
  • Description:
    Tizanidine hydrochloride 100 µg/mL in Acetonitrile
  • Size:
    1 ml
  • Formula:
    C₉H₈ClN₅S
  • Molecular Weight:
    290.17 g/mol
  • Cat. No.:
    77693-438

Specifications

About this item