Tinidazole 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77693-436
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:19387-91-8
- Description:Tinidazole 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₈H₁₃N₃O₄S
- Molecular Weight:247.27 g/mol
- Cat. No.:77693-436