Terbutaline sulfate 100 µg/mL in Acetonitrile:Methanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77693-427
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:23031-32-5
- Description:Terbutaline sulfate 100 µg/mL in Acetonitrile:Methanol
- Size:1 ml
- Formula:C₂₄H₄₀N₂O₁₀S
- Molecular Weight:548.648 g/mol
- Cat. No.:77693-427