Stavudine 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77693-408
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:3056-17-5
- Description:Stavudine 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₀H₁₂N₂O₄
- Molecular Weight:224.22 g/mol
- Cat. No.:77693-408