4-Hydroxybenzoic acid-methyl ester 1000 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-642
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:99-76-3
- Description:4-Hydroxybenzoic acid-methyl ester 1000 µg/mL in Acetonitrile
- Size:1 ml
- Formula:HOC₆H₄CO₂CH₃
- Molecular Weight:152.15 g/mol
- Cat. No.:77691-642