2-Ethylphenol 1000 µg/mL in Methanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-629
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:90-00-6
- Description:2-Ethylphenol 1000 µg/mL in Methanol
- Size:1 ml
- Formula:C₈H₁₀O
- Molecular Weight:122.17 g/mol
- Cat. No.:77691-629