Diclofenac acid 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-604
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:15307-86-5
- Description:Diclofenac acid 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₄H₁₁Cl₂NO₂
- Cat. No.:77691-604