Dichloromethane D2 100 µg/mL in Methanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-601
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:1665-00-5
- Description:Dichloromethane D2 100 µg/mL in Methanol
- Size:1 ml
- Formula:CD₂Cl₂
- Molecular Weight:86.92 g/mol
- Cat. No.:77691-601