2,6-Di-tert-butyl-4-methylphenol 1000 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-598
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:128-37-0
- Description:2,6-Di-tert-butyl-4-methylphenol 1000 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₅H₂₄O
- Molecular Weight:220.35 g/mol
- Cat. No.:77691-598