4,4'-DDT 1000 µg/mL in Toluene, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-593
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:50-29-3
- Description:4,4'-DDT 1000 µg/mL in Toluene
- Size:1 ml
- Formula:C₁₄H₉Cl₅
- Molecular Weight:354.49 g/mol
- Cat. No.:77691-593