4,4'-DDE 1000 µg/mL in Toluene, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-592
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:72-55-9
- Description:4,4'-DDE 1000 µg/mL in Toluene
- Size:1 ml
- Formula:C₁₄H₈Cl₄
- Molecular Weight:318.03 g/mol
- Cat. No.:77691-592