Clarithromycin 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-574
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:81103-11-9
- Description:Clarithromycin 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₃₈H₆₉NO₁₃
- Molecular Weight:747.96 g/mol
- Cat. No.:77691-574