Ciprofloxacin 100 µg/mL in Methanol, Dr. Ehrenstorfer, LGC Standards
77691-571
New Product
:
- CAS No.:85721-33-1
- Description:Ciprofloxacin 100 µg/mL in Methanol
- Size:1 ml
- Formula:C₁₇H₁₈FN₃O₃
- Molecular Weight:331.35 g/mol
- Cat. No.:77691-571