Ciclesonide 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-568
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:126544-47-6
- Description:Ciclesonide 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₃₂H₄₄O₇
- Molecular Weight:540.69 g/mol
- Cat. No.:77691-568