Chlorothiazide 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-564
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:58-94-6
- Description:Chlorothiazide 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₇H₆ClN₃O₄S₂
- Molecular Weight:295.71 g/mol
- Cat. No.:77691-564