Order Entry
Puerto Rico
ContactUsLinkComponent
Chloroparaffin C18 40% Cl 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Chloroparaffin C18 40% Cl 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
  77691-557
New Product
 :  
undefined
Chloroparaffin C18 40% Cl 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
  77691-557
 :  DRE-A11457590CY-100
New Product
 :  

 

  • CAS No.:
    85535-86-0
  • Description:
    Chloroparaffin C18 40% Cl 100 µg/mL in Cyclohexane
  • Size:
    1 ml
  • Cat. No.:
    77691-557