Order Entry
Puerto Rico
ContactUsLinkComponent
 
Chloroparaffin C16 60% Cl 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
  77691-550
New Product
 :  
undefined
Chloroparaffin C16 60% Cl 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
  77691-550
 :  DRE-A11457576CY-100
New Product
 :  

 

  • CAS No.:
    85535-85-9
  • Description:
    Chloroparaffin C16 60% Cl 100 µg/mL in Cyclohexane
  • Size:
    1 ml
  • Cat. No.:
    77691-550