Carazolol 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-526
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:57775-29-8
- Description:Carazolol 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₈H₂₂N₂O₂
- Molecular Weight:298.39 g/mol
- Cat. No.:77691-526