Bisphenol F 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-498
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:620-92-8
- Description:Bisphenol F 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₃H₁₂O₂
- Molecular Weight:200.24 g/mol
- Cat. No.:77691-498