Bezafibrate 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77691-490
New Product
:
- CAS No.:41859-67-0
- Description:Bezafibrate 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₉H₂₀ClNO₄
- Molecular Weight:361.82 g/mol
- Cat. No.:77691-490