Betamethasone 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77691-489
New Product
:
- CAS No.:378-44-9
- Description:Betamethasone 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₂₂H₂₉FO₅
- Molecular Weight:392.47 g/mol
- Cat. No.:77691-489