Azorubin (E122) 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77691-471
New Product
:
- CAS No.:3567-69-9
- Description:Azorubin (E122) 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₂₀H₁₄N₂O₇S₂
- Molecular Weight:502.42 g/mol
- Cat. No.:77691-471